Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rtn3
Entrez Gene ID
394047
Gene Name
reticulon 3
Gene Aliases
wu:fd60d11, zgc:56439, zgc:77630
Chromosome Location
Chr.7: 17716575 - 17753866 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328361.1 NP_001315290.1 3 - 4 511 87
NM_201072.2 NP_957366.1 2 - 3 466 87
XM_021477447.1 XP_021333122.1 4 - 5 3177 87
GenBank mRNA AY555037.1 - 2 - 3 454 87
AY555038.1 - 3 - 4 499 87
BC049315.1 - 2 - 3 454 87
BC062828.1 - 2 - 3 466 87
EH454794.1 - 2 - 3 483 87
EH455188.1 - 3 - 4 508 87
EH521640.1 - 5 - 6 844 87

View Full Product Details