Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hpda
Entrez Gene ID
394142
Gene Name
4-hydroxyphenylpyruvate dioxygenase a
Gene Aliases
fb58f02, hpd, wu:fb58f02, zgc:56326
Chromosome Location
Chr.5: 2814989 - 2852285 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201167.1 NP_957461.1 6 - 7 366 75
XM_005171729.3 XP_005171786.1 6 - 7 402 75
GenBank mRNA AY398361.1 - 5 - 6 330 75
BC046075.1 - 6 - 7 366 75

View Full Product Details