Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc35b2
Entrez Gene ID
402931
Gene Name
solute carrier family 35 member B2
Gene Aliases
papst1, pic, zgc:64182, zgc:77349
Chromosome Location
Chr.20: 51238115 - 51267423 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205635.2 NP_991198.1 4 - 5 845 59
GenBank mRNA BC065347.1 - 4 - 5 819 59
BC090054.1 - 4 - 5 826 59

View Full Product Details