Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
six1b
Entrez Gene ID
404627
Gene Name
SIX homeobox 1b
Gene Aliases
six1, zgc:77345
Chromosome Location
Chr.20: 20499923 - 20501818 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207095.1 NP_996978.1 1 - 2 706 96
GenBank mRNA AY254196.1 - 1 - 2 596 96
AY466110.2 - 1 - 2 672 96
BC066396.1 - 1 - 2 706 96

View Full Product Details