Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gstm.1
Entrez Gene ID
324366
Gene Name
glutathione S-transferase mu, tandem duplicate 1
Gene Aliases
cb760, gstm, wu:fc26g06, zgc:66370
Chromosome Location
Chr.8: 25295234 - 25297928 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212676.1 NP_997841.1 3 - 4 197 72
GenBank mRNA BC057526.1 - 3 - 4 197 72
BC164316.1 - 3 - 4 214 72

View Full Product Details