Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
akirin2
Entrez Gene ID
406855
Gene Name
akirin 2
Gene Aliases
C6orf166, wu:fb16h11, wu:fi04e05, zgc:114001, zgc:55291, zgc:77169
Chromosome Location
Chr.20: 2715944 - 2725787 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213294.2 NP_998459.2 3 - 4 760 79
GenBank mRNA AY394967.1 - 3 - 4 765 79
BC045294.1 - 3 - 4 751 79
BC065319.1 - 3 - 4 689 79
EH552945.1 - 3 - 4 772 79
EH584322.1 - 3 - 4 789 79

View Full Product Details