Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ift20
Entrez Gene ID
414930
Gene Name
intraflagellar transport 20 homolog (Chlamydomonas)
Gene Aliases
zgc:103674
Chromosome Location
Chr.15: 23784748 - 23790172 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001833.2 NP_001001833.2 4 - 5 328 81
XM_005157577.4 XP_005157634.1 3 - 4 414 81
GenBank mRNA AY600453.1 - 2 - 3 251 81
BC081508.1 - 4 - 5 328 81
BC165149.1 - 2 - 3 250 81

View Full Product Details