Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ift20
Entrez Gene ID
414930
Gene Name
intraflagellar transport 20 homolog (Chlamydomonas)
Gene Aliases
zgc:103674
Chromosome Location
Chr.15: 23784748 - 23790172 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001833.2 NP_001001833.2 5 - 6 430 90
XM_005157577.4 XP_005157634.1 4 - 5 90
GenBank mRNA AY600453.1 - 3 - 4 353 90
BC081508.1 - 5 - 6 430 90
BC165149.1 - 3 - 4 352 90

View Full Product Details