Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clic2
Entrez Gene ID
436834
Gene Name
chloride intracellular channel 2
Gene Aliases
zgc:92762
Chromosome Location
Chr.14: 60913 - 70201 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002561.1 NP_001002561.1 2 - 3 313 107
GenBank mRNA BC076239.1 - 2 - 3 313 107

View Full Product Details