Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
paip1
Entrez Gene ID
445110
Gene Name
poly(A) binding protein interacting protein 1
Gene Aliases
fb53g01, wu:fb53g01, zgc:91954
Chromosome Location
Chr.5: 8684652 - 8711046 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003504.1 NP_001003504.1 4 - 5 700 66
GenBank mRNA BC078339.1 - 4 - 5 700 66

View Full Product Details