Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gcc1
Entrez Gene ID
445205
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
zgc:101007
Chromosome Location
Chr.4: 20063256 - 20070035 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003599.2 NP_001003599.1 2 - 3 1269 62
XM_005164735.3 XP_005164792.1 2 - 3 1252 62
XM_021474994.1 XP_021330669.1 3 - 4 1270 62
GenBank mRNA BC078323.1 - 2 - 3 1273 62

View Full Product Details