Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
kop
Entrez Gene ID
493632
Gene Name
askopos
Gene Aliases
zgc:123051
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007787.1 NP_001007788.1 - 966 106
GenBank mRNA AY572416.1 - - 966 106
BC108041.1 - - 1002 106

View Full Product Details