Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tfr1b
Entrez Gene ID
494478
Gene Name
transferrin receptor 1b
Gene Aliases
-
Chromosome Location
Chr.24: 26017094 - 26031250 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009918.2 NP_001009918.2 3 - 4 433 63
GenBank mRNA AY649364.1 - 2 - 3 273 63
BC108025.1 - 3 - 4 433 63

View Full Product Details