Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rrp7a
Entrez Gene ID
550241
Gene Name
ribosomal RNA processing 7 homolog A
Gene Aliases
zgc:110084
Chromosome Location
Chr.3: 1219396 - 1222249 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017579.1 NP_001017579.1 4 - 5 457 61
GenBank mRNA BC092724.1 - 4 - 5 457 61

View Full Product Details