Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccn1l1
Entrez Gene ID
550346
Gene Name
cellular communication network factor 1, like 1
Gene Aliases
Cyr61-l1, cyr61l1, zgc:112060
Chromosome Location
Chr.5: 14113 - 19934 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017653.1 NP_001017653.1 1 - 2 268 67
GenBank mRNA BC093177.1 - 1 - 2 268 67
BC164480.1 - 1 - 2 113 67
GQ273493.1 - 1 - 2 241 67

View Full Product Details