Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
psmb4
Entrez Gene ID
550499
Gene Name
proteasome 20S subunit beta 4
Gene Aliases
im:6909492, zgc:110330
Chromosome Location
Chr.19: 42456922 - 42462486 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017801.1 NP_001017801.1 6 - 7 773 110
XM_005159741.1 XP_005159798.1 6 - 7 110
GenBank mRNA BC092880.1 - 6 - 7 773 110

View Full Product Details