Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gstp2
Entrez Gene ID
553169
Gene Name
glutathione S-transferase pi 2
Gene Aliases
-
Chromosome Location
Chr.14: 3495542 - 3503549 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020513.1 NP_001018349.1 3 - 4 202 75
GenBank mRNA AB194128.1 - 3 - 4 202 75
BC163150.1 - 3 - 4 189 75
BC163154.1 - 3 - 4 189 75

View Full Product Details