Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chrm5a
Entrez Gene ID
553978
Gene Name
cholinergic receptor, muscarinic 5a
Gene Aliases
chrm5, zgc:194865, zgc:194868
Chromosome Location
Chr.17: 463767 - 465239 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020803.1 NP_001018639.1 1 - 1 1053 103
GenBank mRNA BC163412.1 - 1 - 1 1099 103
BC163414.1 - 1 - 1 1099 103

View Full Product Details