Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wnt7aa
Entrez Gene ID
565714
Gene Name
wingless-type MMTV integration site family, member 7Aa
Gene Aliases
wnt7a
Chromosome Location
Chr.11: 27488399 - 27500457 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025540.1 NP_001020711.1 3 - 4 569 88
GenBank mRNA BC163690.1 - 3 - 4 625 88
BC163691.1 - 3 - 4 625 88
DQ074644.1 - 3 - 4 569 88

View Full Product Details