Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wt1b
Entrez Gene ID
568416
Gene Name
WT1 transcription factor b
Gene Aliases
si:ch211-160j6.3, wt1, zgc:152697
Chromosome Location
Chr.18: 45593266 - 45617421 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039634.2 NP_001034723.2 5 - 6 938 63
XM_005159243.4 XP_005159300.1 5 - 6 1211 63
GenBank mRNA BC124087.1 - 5 - 6 938 63
BC164718.1 - 5 - 6 796 63
DQ145942.1 - 5 - 6 762 63

View Full Product Details