Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
asb15b
Entrez Gene ID
568579
Gene Name
ankyrin repeat and SOCS box containing 15b
Gene Aliases
si:dkey-3n7.3
Chromosome Location
Chr.4: 4120570 - 4129187 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039890.1 NP_001034979.1 1 - 2 108 102

View Full Product Details