Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tiparp
Entrez Gene ID
563506
Gene Name
TCDD-inducible poly(ADP-ribose) polymerase
Gene Aliases
fc03g05, sb:cb841, si:dkey-33k14.4, wu:fc03g05, zgc:153056
Chromosome Location
Chr.18: 200153 - 234060 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045019.3 NP_001038484.1 2 - 3 1000 63
GenBank mRNA CA472479.1 - 1 - 2 119 63
EE311950.1 - 1 - 2 220 63
EE316647.1 - 1 - 2 509 63

View Full Product Details