Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tp53i11b
Entrez Gene ID
556618
Gene Name
tumor protein p53 inducible protein 11b
Gene Aliases
fi47b03, si:ch211-122k23.2, tp53i11, wu:fi47b03, zgc:153054
Chromosome Location
Chr.18: 27439843 - 27569316 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077249.1 NP_001070717.1 6 - 7 624 70
XM_021467658.1 XP_021323333.1 6 - 7 633 70
GenBank mRNA BC124630.1 - 6 - 7 624 70
BC164984.1 - 4 - 5 362 70

View Full Product Details