Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc29a4
Entrez Gene ID
559668
Gene Name
solute carrier family 29 member 4
Gene Aliases
zgc:158679
Chromosome Location
Chr.3: 40315383 - 40349735 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080572.2 NP_001074041.2 8 - 9 1272 69
NM_001362610.1 NP_001349539.1 8 - 9 1281 69
GenBank mRNA BC129376.1 - 8 - 9 1272 69

View Full Product Details