Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chrd
Entrez Gene ID
30161
Gene Name
chordin
Gene Aliases
chd, din, fe47d04, wu:fe47d04
Chromosome Location
Chr.15: 46791702 - 46828902 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130973.3 NP_571048.2 3 - 4 511 61
GenBank mRNA AF034606.1 - 3 - 4 513 61

View Full Product Details