Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ephb4b
Entrez Gene ID
30691
Gene Name
eph receptor B4b
Gene Aliases
EphB-rtk8, cb417, rtk8
Chromosome Location
Chr.23: 44349830 - 44400916 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131417.1 NP_571492.1 4 - 5 824 84
GenBank mRNA AJ005029.1 - 4 - 5 824 84
BC163275.1 - 4 - 5 906 84
KM655609.1 - 4 - 5 824 84

View Full Product Details