Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
odc1
Entrez Gene ID
114426
Gene Name
ornithine decarboxylase 1
Gene Aliases
CHUNP6922, fc54f04, fi06d08, odc, wu:fc54f04, wu:fi06d08
Chromosome Location
Chr.17: 51743921 - 51757548 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131801.2 NP_571876.1 7 - 8 1053 71
GenBank mRNA AB069857.1 - 7 - 8 1053 71
AF290981.1 - 6 - 7 918 71
BC047796.1 - 6 - 7 1018 71
BC063932.1 - 7 - 8 1024 71
BC097034.1 - 6 - 7 1131 71
BC165342.1 - 5 - 6 790 71

View Full Product Details