Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gpc4
Entrez Gene ID
118437
Gene Name
glypican 4
Gene Aliases
chunp6920, fc47a08, fe05f10, gpc4,, gpc4/6, gpc6, kny, knypek, wu:fc47a08, wu:fe05f10, zgc:194854
Chromosome Location
Chr.14: 31092327 - 31151148 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131860.2 NP_571935.2 3 - 4 802 79
GenBank mRNA BC163403.1 - 3 - 4 802 79

View Full Product Details