Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sf3b4
Entrez Gene ID
192318
Gene Name
splicing factor 3b, subunit 4
Gene Aliases
chunp6902, fb36a04, wu:fb36a04
Chromosome Location
Chr.19: 7424358 - 7430856 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153661.4 NP_705947.3 1 - 2 62 85
GenBank mRNA BC056532.1 - 1 - 2 48 85
BC067655.1 - 1 - 2 52 85
EH501773.1 - 1 - 2 84 85

View Full Product Details