Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arntl1b
Entrez Gene ID
352935
Gene Name
aryl hydrocarbon receptor nuclear translocator-like 1b
Gene Aliases
bmal1b, zfBMAL3
Chromosome Location
Chr.7: 209733 - 240518 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178300.1 NP_840085.1 14 - 15 1514 72
XM_005166616.4 XP_005166673.1 17 - 18 1772 72
XM_009303573.3 XP_009301848.1 16 - 17 1703 72
XM_021477412.1 XP_021333087.1 16 - 17 1710 72
GenBank mRNA AB087257.1 - 14 - 15 1514 72
BC134895.1 - 17 - 18 1757 72

View Full Product Details