Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pvalb8
Entrez Gene ID
360208
Gene Name
parvalbumin 8
Gene Aliases
ocma, pvalb3a, zgc:101834
Chromosome Location
Chr.3: 61154219 - 61162750 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182937.2 NP_891982.2 3 - 4 463 104
GenBank mRNA AF425739.1 - 3 - 4 254 104
BC085451.1 - 3 - 4 463 104
BC164552.1 - 2 - 3 229 104

View Full Product Details