Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
evla
Entrez Gene ID
58003
Gene Name
Enah/Vasp-like a
Gene Aliases
Evl, id:ibd2260, zgc:65883, zgc:77388
Chromosome Location
Chr.17: 30900495 - 30976007 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199519.3 NP_955813.3 10 - 11 1405 67
XM_005158792.4 XP_005158849.1 11 - 12 2031 67
XM_005158793.4 XP_005158850.1 11 - 12 2024 67
XM_005158794.4 XP_005158851.1 11 - 12 1819 67
XM_005158795.4 XP_005158852.1 11 - 12 2158 67
XM_005158796.4 XP_005158853.1 9 - 10 1735 67
XM_005158797.4 XP_005158854.1 10 - 11 1272 67
XM_005158798.4 XP_005158855.1 10 - 11 1270 67
XM_005158799.4 XP_005158856.1 10 - 11 1267 67
XM_021467223.1 XP_021322898.1 11 - 12 2029 67
GenBank mRNA BC056325.1 - 10 - 11 1376 67
BC065353.1 - 10 - 11 1369 67

View Full Product Details