Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cul3a
Entrez Gene ID
324655
Gene Name
cullin 3a
Gene Aliases
cul3, wu:fc37e01
Chromosome Location
Chr.2: 47644041 - 47681461 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199691.2 NP_955985.2 15 - 16 2324 130
GenBank mRNA AY423034.1 - 15 - 16 2317 130
BC060901.1 - 15 - 16 2266 130
BC065357.1 - 15 - 16 2286 130
DT057506.1 - 4 - 5 555 130

View Full Product Details