Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lmbrd2b
Entrez Gene ID
335257
Gene Name
LMBR1 domain containing 2b
Gene Aliases
fk97a10, wu:fk97a10, zgc:73387
Chromosome Location
Chr.21: 22400962 - 22422876 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199944.1 NP_956238.1 17 - 18 2133 68
GenBank mRNA BC063324.1 - 17 - 18 2133 68

View Full Product Details