Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slmapb
Entrez Gene ID
393146
Gene Name
sarcolemma associated protein b
Gene Aliases
slmap, zgc:55817
Chromosome Location
Chr.23: 19589821 - 19604413 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200177.1 NP_956471.1 3 - 4 229 120
XM_005162050.4 XP_005162107.1 3 - 4 120
GenBank mRNA BC045462.1 - 3 - 4 229 120
BC165867.1 - 2 - 3 209 120

View Full Product Details