Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ubtfl
Entrez Gene ID
393978
Gene Name
upstream binding transcription factor, like
Gene Aliases
Tcfubf, zgc:63557
Chromosome Location
Chr.2: 32357606 - 32388253 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201003.1 NP_957297.1 13 - 14 1480 75
XM_005163486.3 XP_005163543.1 13 - 14 75
XM_005163487.3 XP_005163544.1 13 - 14 75
XM_005163488.3 XP_005163545.1 13 - 14 75
XM_005163489.3 XP_005163546.1 13 - 14 75
XM_005163490.3 XP_005163547.1 13 - 14 75
XM_009298612.2 XP_009296887.1 13 - 14 75
XM_009298613.2 XP_009296888.1 13 - 14 75
XM_009298614.2 XP_009296889.1 13 - 14 75
XM_017357972.2 XP_017213461.1 12 - 13 75
GenBank mRNA BC055140.1 - 13 - 14 1480 75

View Full Product Details