Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps3
Entrez Gene ID
336550
Gene Name
ribosomal protein S3
Gene Aliases
fb13d09, wu:fb13d09, zgc:56088
Chromosome Location
Chr.18: 3038173 - 3049799 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201153.1 NP_957447.1 3 - 4 286 97
GenBank mRNA AY394937.1 - 3 - 4 97
BC045902.1 - 3 - 4 286 97
BC165099.1 - 3 - 4 293 97

View Full Product Details