Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col1a1b
Entrez Gene ID
325675
Gene Name
collagen, type I, alpha 1b
Gene Aliases
alpha3(I), col1a3, fj59a10, hm:zeh0348, wu:fa95h05, wu:fd02a10, wu:fj59a10
Chromosome Location
Chr.12: 3095617 - 3133500 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201478.1 NP_958886.1 7 - 8 644 138
XM_005155976.4 XP_005156033.1 7 - 8 687 138
XM_005155980.4 XP_005156037.1 7 - 8 690 138
GenBank mRNA BC058045.1 - 7 - 8 644 138

View Full Product Details