Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scamp2
Entrez Gene ID
406687
Gene Name
secretory carrier membrane protein 2
Gene Aliases
wu:fe48e09, zgc:56290, zgc:77037
Chromosome Location
Chr.25: 3015674 - 3034664 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213378.2 NP_998543.1 7 - 8 825 135
GenBank mRNA BC066575.1 - 7 - 8 785 134
EH539419.1 - 7 - 8 753 135

View Full Product Details