Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mospd1
Entrez Gene ID
406678
Gene Name
motile sperm domain containing 1
Gene Aliases
wu:fc26f10, zgc:56336
Chromosome Location
Chr.14: 106951 - 118856 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213369.1 NP_998534.1 4 - 5 624 97
GenBank mRNA BC046085.1 - 4 - 5 624 97

View Full Product Details