Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mpv17l2
Entrez Gene ID
436840
Gene Name
MPV17 mitochondrial inner membrane protein like 2
Gene Aliases
zgc:103505, zgc:92754
Chromosome Location
Chr.2: 182410 - 190861 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002567.1 NP_001002567.1 2 - 3 703 94
GenBank mRNA BC076231.1 - 2 - 3 703 94
BC086824.1 - 2 - 3 682 94

View Full Product Details