Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aarsd1
Entrez Gene ID
445178
Gene Name
alanyl-tRNA synthetase domain containing 1
Gene Aliases
im:7152658, zgc:101066
Chromosome Location
Chr.12: 6049358 - 6063311 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003572.1 NP_001003572.1 11 - 12 1166 101
GenBank mRNA BC077118.1 - 11 - 12 1166 101

View Full Product Details