Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
syt9b
Entrez Gene ID
445476
Gene Name
synaptotagmin IXb
Gene Aliases
zgc:91875
Chromosome Location
Chr.25: 15841719 - 15905903 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003985.1 NP_001003985.1 2 - 3 731 68
GenBank mRNA BC080256.1 - 2 - 3 731 68
BC163971.1 - 2 - 3 550 68

View Full Product Details