Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cltca
Entrez Gene ID
323579
Gene Name
clathrin, heavy chain a (Hc)
Gene Aliases
cb1033, cltc, wu:fc03e11, wu:fc49a11, wu:fd07f02
Chromosome Location
Chr.10: 50403 - 108768 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005391.2 NP_001005391.2 2 - 3 530 87
XM_009305358.3 XP_009303633.1 2 - 3 520 87
GenBank mRNA AY648777.1 - 2 - 3 467 87
BC163714.1 - 2 - 3 278 87

View Full Product Details