Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tspan7b
Entrez Gene ID
449539
Gene Name
tetraspanin 7b
Gene Aliases
tm4sf2b, wu:fb53h11, zgc:92829
Chromosome Location
Chr.22: 11440705 - 11493236 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130818.1 NP_001124290.1 6 - 7 734 71
GenBank mRNA BC081682.1 - 6 - 7 685 71

View Full Product Details