Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
il12ba
Entrez Gene ID
445373
Gene Name
interleukin 12Ba
Gene Aliases
IL-12 p40, il12b, il12p40.b, zgc:152789
Chromosome Location
Chr.14: 34951202 - 34958499 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007108.1 NP_001007109.1 4 - 5 530 70
GenBank mRNA AB183002.1 - 4 - 5 530 70
BC121764.1 - 4 - 5 529 70
BC164577.1 - 3 - 4 501 70

View Full Product Details