Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
usp2a
Entrez Gene ID
494031
Gene Name
ubiquitin specific peptidase 2a
Gene Aliases
zgc:92134
Chromosome Location
Chr.15: 20900627 - 20939599 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008574.2 NP_001008574.2 11 - 12 1385 115
XM_005157513.4 XP_005157570.1 13 - 14 1888 115
XM_005157514.4 XP_005157571.1 13 - 14 1982 115
XM_005157515.4 XP_005157572.1 13 - 14 1880 115
XM_005157516.4 XP_005157573.1 11 - 12 1456 115
XM_017350951.2 XP_017206440.1 13 - 14 1883 115
XM_021481396.1 XP_021337071.1 13 - 14 1986 115
GenBank mRNA BC086963.1 - 11 - 12 1226 115

View Full Product Details