Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
adoa
Entrez Gene ID
494091
Gene Name
2-aminoethanethiol (cysteamine) dioxygenase a
Gene Aliases
wu:fc32g01, wu:fc52c12, zgc:101580
Chromosome Location
Chr.12: 8474868 - 8476513 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008634.1 NP_001008634.1 1 - 1 848 63
GenBank mRNA BC086706.1 - 1 - 1 848 63

View Full Product Details