Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ogna
Entrez Gene ID
541445
Gene Name
osteoglycin, paralog a
Gene Aliases
ogn, si:ch211-103f16.4, zgc:113545
Chromosome Location
Chr.22: 10454603 - 10459880 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013570.2 NP_001013588.1 2 - 3 334 143
GenBank mRNA BC090507.1 - 2 - 3 338 143

View Full Product Details