Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hmgcrb
Entrez Gene ID
541479
Gene Name
3-hydroxy-3-methylglutaryl-CoA reductase b
Gene Aliases
hmgcr2, im:6912716, zgc:110746
Chromosome Location
Chr.21: 2506024 - 2533246 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014292.2 NP_001014314.1 14 - 15 1941 82
GenBank mRNA BC091792.1 - 14 - 15 1945 82

View Full Product Details